▸
Tubes
▸
Test Tubes
▸
Reusable Test Tubes
▸
Soda Glass Test tubes
Actual product may vary
Fisherbrand
Soda Glass Test tubes
Use these test tubes for general mixing and simple laboratory work. Soda Glass Test tubes will withstand moderate warming (ex. in a water bath).
Have Questions?
Catalog Number
Diameter (Metric)
Length (Metric)
FKS12319329
10 mm
50 mm
Catalog number
FKS12319329
Price (SGD)
186.00
Pack
Add to cart
Diameter (Metric):
10 mm
Length (Metric):
50 mm
Price (SGD)
186.00
Pack
Add to cart
Light walled test tube with rim
Should not be placed directly into naked flames
Available in packs of 250
Use these test tubes for general mixing and simple laboratory work. Soda Glass Test tubes will withstand moderate warming (ex. in a water bath).
Specifications
Certifications/Compliance
ISO 4142:2002
Length (Metric)
50 mm
For Use With
General mixing, Simple laboratory work
Includes
Rim
Autoclavable
Not Autoclavable
Diameter (Metric)
10 mm