Test Tube Brushes with Cotton Yarn Tip
Test Tube Brushes with Cotton Yarn Tip
Actual product may vary
Fisherbrand
Test Tube Brushes with Cotton Yarn Tip
Features With cotton yarn tip. Pinnacle Brush™ Test Tube Brushes with Cotton Yarn Tip have a galvanised steel wire handle.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) Bristle
FKS1231833825 mm
FKS1230833815 mm
Catalog number FKS12318338
Price (SGD)
67.00
PK
Add to cart
Diameter (Metric) Bristle:
25 mm
Price (SGD)
67.00
PK
Add to cart
  • Cotton yarn tip
  • Metal handle
Specifications
Product TypeTest Tube Brush
Bristle MaterialFiber
Handle MaterialMetal
Diameter (Metric)3.5 cm
Diameter (Metric) Bristle25 mm
For Use With (Equipment)Test tubes
Length (Metric)22 cm
Length (Metric) Brush7 cm
Length (Metric) Handle15 cm
Quantity10 Pk.
SizeBrush Circumference: 11 cm
Unit SizePK