Check Order Status
Cork Stoppers
Actual product may vary
Fisherbrand
Cork Stoppers
For use in education, homebrewing and arts and crafts industries.
Have Questions?
Catalog Number
Diameter (Metric) Bottom
Diameter (Metric) Top
FKS12316467
21 mm
24 mm
Catalog number
FKS12316467
Price (SGD)
14.00
PK
Add to cart
Diameter (Metric) Bottom:
21 mm
Diameter (Metric) Top:
24 mm
Price (SGD)
14.00
PK
Add to cart
Available in different sizes
Specifications
Length (Metric)
32 mm
Quantity
10/Pk.
Diameter (Metric) Bottom
21 mm
Diameter (Metric) Top
24 mm
Material
Cork
Product Type
Cork Stopper