KECK™ Assembly Set
KECK™ Assembly Set
Actual product may vary
KECK™ Assembly Set
KECK™ Assembly Set
Fisherbrand
KECK™ Assembly Set
The KECK™ assembly set serves as a connection system for a suction bottle, via which the vacuum pump can be connected.
Have Questions?
Catalog NumberSize
FKS123138969 mm
Catalog number FKS12313896
Price (SGD)
272.00
PK
Add to cart
Size:
9 mm
Price (SGD)
272.00
PK
Add to cart
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Buechner funnels are cylinders with an integrated large-pored sieve plate. Without additional filter paper, only coarse filtration is possible. Buechner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.
  • With removable plastic hose connection (PBT), short and long screw (PP) and seals (VMQ, EPDM)
  • Suitable for filtering flasks 100 - 20000 ml
Specifications
Quantity10 Pk.
MaterialPBT, Polypropylene, VMQ, EPDM
For Use With (Application)Filtering; Vacuum filtration; Safe working under vacuum
Size9 mm
Unit SizePK