SuperFrost Plus™ Adhesion slides
Product Image
Fisherbrand
SuperFrost Plus™ Adhesion slides
Minimizes tissue loss during staining resulting in quick turn-around times and reduced risk associated with tissue loss and repeat testing. Epredia™ SuperFrost Plus Slides secure frozen and formalin-fixed, paraffin embedded tissue sections as well as cytology preparations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorCorner Style
FKS12312148YellowTraditional
FKS10149870WhiteTraditional
Catalog number FKS12312148
Price (SGD)
92.00
PK
Add to cart
Color:
Yellow
Corner Style:
Traditional
Price (SGD)
92.00
PK
Add to cart
  • No time wasted on slide-coating procedures
  • No money wasted on expensive adhesives
  • Reliable and uniform coating
  • Minimal sample loss
  • Reliable and safe adhesive action
Specifications
Edges90°, Ground
Length (Metric)75 mm
TypeAdhesion Slide
Width (Metric)25 mm
ColorYellow
Corner StyleTraditional
Quantity72/Pk.
Unit SizePK