Porcelain Evaporating Basins
Porcelain Evaporating Basins
Actual product may vary
Fisherbrand
Porcelain Evaporating Basins
Use these dishes for a variety of applications. Haldenwanger™ Porcelain Basins feature a smooth internal finish, constructed from optimized laboratory porcelain.
Have Questions?
Catalog NumberCapacity (Metric)Height (Metric)
FKS12310699140 mL40 mm
Catalog number FKS12310699
Price (SGD)
152.00
EA
Add to cart
Capacity (Metric):
140 mL
Height (Metric):
40 mm
Price (SGD)
152.00
EA
Add to cart
  • Shallow form, round-bottom porcelain basin
  • Available in a variety of sizes
  • Sizes up to 175mL glazed except rim
  • 1,000mL and larger are glazed except outside base
  • Includes spout for easy pouring
  • Complies with DIN VDE 0335, type C 100
Specifications
Capacity (Metric)140 mL
MaterialPorcelain
ShapeRound
TypeEvaporating Basin
Height (Metric)40 mm
Diameter (Metric)97 mm
Unit SizeEA