Check Order Status
B-Safe Biohazard Bags
Actual product may vary
Fisherbrand
B-Safe Biohazard Bags
Use Alpha Packaging™ B-Safe Biohazard Bags for heavy duty biohazardous waste. The bags are yellow with biohazard printed on them.
Have Questions?
Catalog Number
Color
FKS12310609
Yellow
Catalog number
FKS12310609
Price (SGD)
335.00
PK
Add to cart
Color:
Yellow
Price (SGD)
335.00
PK
Add to cart
Bench top dispenser boxes
Specifications
Color
Yellow
For Use With (Application)
Waste Disposal
Material
Polyethylene
Thickness (English)
5 mil
Dimensions (L x W)
760 x 455 mm
Product Type
Waste Disposal Bags