HDPE, Round Aspirator Bottle
HDPE, Round Aspirator Bottle
Actual product may vary
Fisherbrand
HDPE, Round Aspirator Bottle
Capacity Metric: 25 L
Have Questions?
Catalog number FKS12310459
Price (SGD)
734.00
EA
Add to cart
Price (SGD)
734.00
EA
Add to cart
  • Air-vented 12mm bore tap
  • Screw on cap
  • Plastic covered band around shoulder includes integral lifting handles
Specifications
Capacity (English)6.6 gal.
Capacity (Metric)25 L
Diameter (Metric) Outer315 mm
Height (Metric) With Closure414 mm
IncludesClosure, Handles, 12mm bore tap spigot
MaterialHDPE
Neck StyleNarrow
Product TypeAspirator Bottle
ShapeCylindrical
Unit SizeEA