Wash Bottle, Deionized Water
Wash Bottle, Deionized Water
Actual product may vary
Fisherbrand
Wash Bottle, Deionized Water
Right-to-Know Safety-Labeled wash bottles.
Have Questions?
Catalog NumberCapacity (Metric)
FKS12307248500 mL
Catalog number FKS12307248
Price (SGD)
140.00
EA
Add to cart
Capacity (Metric):
500 mL
Price (SGD)
140.00
EA
Add to cart
  • Wide mouth of 53 mm (2 in.) for easy refilling
  • Pre-printed with Chemical name and formula, National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Translucent LDPE bottles
  • Color-coded polypropylene closure
Specifications
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorTranslucent, Blue
IncludesPack of 4 each of “DI Water” wash botttles
MaterialLDPE
Quantity4/Pk.
SterilityNon-sterile
Closure ColorBlue
Closure IncludedYes
AutoclavableNot Autoclavable
DisposableReusable
GraduatedNo
MouthWide, Threaded
ShapeCylindrical
Product TypeWash Bottle
Chemical LabelDeionized Water