Polypropylene Cleanroom Face Masks
Polypropylene Cleanroom Face Masks
Actual product may vary
Fisherbrand
Polypropylene Cleanroom Face Masks
Disposable, low-particulate-generating masks for Class 100 cleanrooms. Berkshire International™ Polypropylene Clean Room Face Masks are constructed of three layers of lint-free material.
Have Questions?
Catalog NumberSizeWidth (Metric)
FKS12306347Standard180 mm
Catalog number FKS12306347
Price (SGD)
-
Size:
Standard
Width (Metric):
180 mm
  • Disposable masks specifically designed for Class 100 cleanrooms.
  • Very low particulate generation.
  • Filtration efficiency at 1.0mm is 96%.
  • Three layers of lint-free material.
  • Two outer layers are made from lint-free, wet laid, non-woven material and the centre filtration medium is made of fine, melt blown polypropylene.
  • Each mask has a metal nose piece that is easy to bend, providing a contoured comfortable fit.
  • Folded portion of the mask extends to cover chin area.
  • Individually packed.
  • Dispenser can be usefully stationed in the changing area of any cleanroom.
Specifications
Product TypeCleanroom Face Mask
MaterialPolypropylene
SizeStandard
Cleanroom ClassClass 10 (ISO 4)
ModelBCRM
Packaging Quantity50/Pk
Width (Metric)180 mm
Unit SizePK