Finely Ground Edge Glass Microscopic Slides
Finely Ground Edge Glass Microscopic Slides
Actual product may vary
Fisherbrand
Finely Ground Edge Glass Microscopic Slides
Packed with tissue to protect glass surfaces. Scientific Glass Labs™ Finely Ground Edge Glass Microscope Slides are best used with large sections.
Have Questions?
Catalog NumberWidth (Metric)Dimensions (L x W)
FKS1230215852 mm76 x 52 mm
Catalog number FKS12302158
Price (SGD)
240.00
PK
Add to cart
Width (Metric):
52 mm
Dimensions (L x W):
76 x 52 mm
Price (SGD)
240.00
PK
Add to cart
With finely ground edges. Packed with tissue to protect glass surfaces. For use with large sections.
Specifications
EdgesFinely Ground
Length (Metric)76 mm
Thickness (Metric)1.2 to 1.5 mm
Width (Metric)52 mm
Quantity50 Pack
Dimensions (L x W)76 x 52 mm
Unit SizePK