PYREX™ Wide Mouth Heavy-Duty Erlenmeyer Flasks
PYREX™ Wide Mouth Heavy-Duty Erlenmeyer Flasks
Actual product may vary
Fisherbrand
PYREX™ Wide Mouth Heavy-Duty Erlenmeyer Flasks
Use these flasks for a variety of applications. PYREX™ Wide Neck Heavy-Duty Erlenmeyer Flasks are designed for longer life and feature a high resistance to chemical attack, rugged tooled top finish and wide mouth.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Diameter (Metric) OuterGraduation Range
FKS122014081 L29.2 cm168 mm250 to 1000 mL
FIS10-090B250 mL18.4 cm106 mm50 to 200 mL
FIS10-090C500 mL23.8 cm137 mm100 to 500 mL
FIS10-090A125 mL14.6 cm84 mm50 to 125 mL
FIS10-090E2000 mL37.2 cm198 mm400 to 1800 mL
FKS106420912000 mL37.2 cm198 mm400 to 1800 mL
Catalog number FKS12201408
Price (SGD)
531.00
PK
Add to cart
Capacity (Metric):
1 L
Height (Metric):
29.2 cm
Diameter (Metric) Outer:
168 mm
Graduation Range:
250 to 1000 mL
Price (SGD)
531.00
PK
Add to cart
Use these flasks for a variety of applications. PYREX™ Wide Neck Heavy-Duty Erlenmeyer Flasks are designed for longer life and feature a high resistance to chemical attack, rugged tooled top finish and wide mouth.
A wide mouth flask especially recommended for use as a titration flask. For convenience, these flasks are graduated to show the approximate capacity and have an extra large marking spot.
Specifications
DescriptionWhite Ceramic Enamel scale graduations; Top style: Heavy duty rim
Diameter (English) Outer6.6 in.
Capacity (Metric)1 L
MaterialBorosilicate Glass, Borosilicate Glass
Graduation Range250 to 1000 mL
Graduations50 mL
Height (English)11.4 in.
Height (Metric)29.2 cm
Neck StyleWide Mouth
No. per Case24/Cs.
No. per Pack6/Pk.
ShapeConical
Writing SurfaceExtra Large White Square Marking
DisposableReusable
Mouth StyleWide
SterilityNon-sterile
Diameter (Metric) Outer168 mm
Closure Size11
Unit SizePK