Economical choice since only about 20mL of buffer is used per calibration. Hamilton™ DuraCal™ 10.01 Value pH Buffers have actual values determined by a DKD laboratory, accredited for pH measurement.
Have Questions?
Catalog Number
Quantity
FKS12201405
500 mL
Catalog number FKS12201405
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
pH10.01
Quantity500 mL
Unit SizePK
10.01 pH value
Immune to micro-organisms
Expiration date on the bottle
First class certificate with traceability to international standards