LDPE Multi-lingual Printed Wash Bottles
LDPE Multi-lingual Printed Wash Bottles
Actual product may vary
Fisherbrand
LDPE Multi-lingual Printed Wash Bottles
Imprinted with Globally Harmonized Symbols, NFPA diamond, risk phrases and safety information. Azlon™ LDPE Multi-lingual Printed Wash Bottles display chemical names and key safety information in four European languages, eliminating the need for multiple wash bottles across European language speaking countries.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Chemical LabelDescription
FKS12186220250 mLIsopropanolIndelibly printed in English, French, German and Spanish
FKS11966815500 mLAcetoneIndelibly printed in English, French, German and Spanish
FKS10233610500 mLMethanolIndelibly printed in English, French, German and Spanish
FKS10690121500 mLIsopropanolIndelibly printed in English, French, German and Spanish
FKS10203370500 mLDeionized WaterIndelibly printed in English, French, German and Spanish
FKS10191740500 mLDistilled WaterIndelibly printed in English, French, German and Spanish
Catalog number FKS12186220
Price (SGD)
133.00
PK
Add to cart
Capacity (Metric):
250 mL
Chemical Label:
Isopropanol
Description:
Indelibly printed in English, French, German and Spanish
Price (SGD)
133.00
PK
Add to cart
Imprinted with Globally Harmonized Symbols, NFPA diamond, risk phrases and safety information. Azlon™ LDPE Multi-lingual Printed Wash Bottles display chemical names and key safety information in four European languages, eliminating the need for multiple wash bottles across European language speaking countries.
  • Multi-lingual wide neck wash bottles
  • Indelibly printed
  • Fitted with color-coded, one-piece PP closures that maintain their shape in use
  • Closures feature precision jet-tip aperture for fine control
  • Available in three water and five solvent labels
Specifications
Capacity (Metric)250 mL
Capacity (English)8.45 oz.
ColorTranslucent
DescriptionIndelibly printed in English, French, German and Spanish
IncludesBlue Polypropylene closure
MaterialLDPE
Quantity5/PK
Temperature Range (Metric)–50 to 80°C
Closure ColorBlue
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthWide
ShapeCylindrical
Height (Metric)140 mm
Product TypeMulti-Lingual Printed Wash Bottle
Chemical LabelIsopropanol
Closure MaterialPolypropylene
Diameter (Metric)60 mm
Closure TypeNon-venting Cap
Through the Closure DispensingYes
Unit SizePK