Polypropylene Analytical Funnels
Polypropylene Analytical Funnels
Actual product may vary
Fisherbrand
Polypropylene Analytical Funnels
Manufactured with polypropylene for strength and durability. Kartell™ Polypropylene Analytical Funnels are designed specifically for analytical chemistry.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric)TopHeight (Metric)Length (Metric) Stem
FKS1217778020 mL46mm79 mm43mm
FKS1192984850 mL66mm112 mm62mm
FKS1191984810 mL37mm63.4 mm37mm
Catalog number FKS12177780
Price (SGD)
115.00
PK
Add to cart
Capacity (Metric):
20 mL
Diameter (Metric)Top:
46mm
Height (Metric):
79 mm
Length (Metric) Stem:
43mm
Price (SGD)
115.00
PK
Add to cart
Manufactured with polypropylene for strength and durability. Kartell™ Polypropylene Analytical Funnels are designed specifically for analytical chemistry.
  • Takes standard size filter paper
  • 60° angled sides with internal and external ribs to speed filtration and avoid pressure lock
  • Withstands temperatures up to 135°C for short periods
  • Autoclavable at 121°C for 20min
  • Suitable for foodstuff
Specifications
Diameter (Metric)Top46mm
Capacity (Metric)20 mL
MaterialPolypropylene
Height (Metric)79 mm
AutoclavableAutoclavable
For Use With (Application)Suitable for foodstuff
Length (Metric) Stem43mm
Max. Temperature (Metric)135°C
Quantity20/Pk.
Unit SizePK