Check Order Status
Translucent Polypropylene Y-shape Tubing Connector
Actual product may vary
Fisherbrand
Translucent Polypropylene Y-shape Tubing Connector
Constructed of translucent polypropylene. Accommodates three tubes with Y-shaped stem and arms.
Have Questions?
Change view
Catalog Number
Tubing Size
FKS12110062
4 to 5 mm tubing I.D.
FKS11833930
3 mm tubing I.D.
FKS11843970
8 to 9 mm tubing I.D.
3 Options
Catalog number
FKS12110062
Price (SGD)
112.00
PK
Add to cart
Tubing Size:
4 to 5 mm tubing I.D.
Price (SGD)
112.00
PK
Add to cart
Translucent polypropylene construction
Accommodates 3 tubes of the same diameter
Specifications
Material
Polypropylene
Overall Length (Metric)
35 mm
Tubing Size
4 to 5 mm tubing I.D.
Width (Metric)
21 mm
Shape
Y
Color
Translucent
Product Type
Tubing Connector