Azlon™ Polypropylene Spatula
Azlon™ Polypropylene Spatula
Actual product may vary
Fisherbrand
Azlon™ Polypropylene Spatula
Manufactured from polypropylene. SciLabWare™ Azlon™ Polypropylene Spatula is virtually unbreakable in normal use and retain shape after flexing.
Have Questions?
Catalog NumberLength (Metric)
FKS12099867200 mm
Catalog number FKS12099867
Price (SGD)
-
Length (Metric):
200 mm
Manufactured from polypropylene. SciLabWare™ Azlon™ Polypropylene Spatula is virtually unbreakable in normal use and retain shape after flexing.
  • Molded strong tapered spatulas with chamfered ends
  • Virtually unbreakable in normal use and retain shape after flexing
  • Good chemical and heat resistance
  • Autoclavable at 121°C
Specifications
AutoclavableAutoclavable
Product TypeSpatula
Width (Metric)28 mm
Dimensions (L x W)200 x 28 mm
MaterialPolypropylene
Length (Metric)200 mm
Unit SizePK