Quickfit™ Borosilicate Glass Still Head Adapter
Quickfit™ Borosilicate Glass Still Head Adapter
Actual product may vary
Fisherbrand
Quickfit™ Borosilicate Glass Still Head Adapter
Connect a flask with a condenser in a distillation assembly. The QuickFit™ still head adapter is designed to accept a thermometer as well.
Have Questions?
Catalog NumberStandard Taper Joint
FKS12098483Cone Size: 29/32 (Condenser), 29/32 (flask); Socket Size: 14/23
Catalog number FKS12098483
Price (SGD)
290.00
EA
Add to cart
Standard Taper Joint:
Cone Size: 29/32 (Condenser), 29/32 (flask); Socket Size: 14/23
Price (SGD)
290.00
EA
Add to cart
Connect a flask with a condenser in a distillation assembly. The QuickFit™ still head adapter is designed to accept a thermometer as well.
  • Used in distillation assemblies for connecting flasks to condenser
  • Upper socket designed to accept thermometer either via cone/screw thread adapter or thermometer pocket
Specifications
For Use With (Application)Connecting flask and condenser
TypeStill Head Adapter
MaterialBorosilicate Glass
Standard Taper JointCone Size: 29/32 (Condenser), 29/32 (flask); Socket Size: 14/23
Unit SizeEA