Plastilab Untapered T-Shaped Tubing Connectors
Plastilab Untapered T-Shaped Tubing Connectors
Actual product may vary
Plastilab Untapered T-Shaped Tubing Connectors
Plastilab Untapered T-Shaped Tubing Connectors
Fisherbrand
Plastilab Untapered T-Shaped Tubing Connectors
Designed with serrations to hold hoses tightly and prevent slipping.
Have Questions?
Catalog NumberPort SizePackaging Quantity
FKS120905826.3mm bore10/Pk.
Catalog number FKS12090582
Price (SGD)
30.00
PK
Add to cart
Port Size:
6.3mm bore
Packaging Quantity:
10/Pk.
Price (SGD)
30.00
PK
Add to cart
Designed with serrations to hold hoses tightly and prevent slipping.
  • Made from polypropylene
  • Autoclavable
  • Chemical- and temperature-resistant
Specifications
Certifications/ComplianceMeet with the Food and Drug regulations. 90/128/CEE 92/39/CEE e FDA 21 CFR 177.1520, CE and IVD
Diameter (Metric)10 mm
AutoclavableAutoclavable
MaterialPolypropylene
For Use With (Application)Suitable for foodstuff
Packaging Quantity10/Pk.
Port Size6.3mm bore
ShapeT-shape
Product TypeUntapered T Tubing Connector
Unit SizePK