Multipurpose Specimen Storage Containers
Multipurpose Specimen Storage Containers
Actual product may vary
Fisherbrand
Multipurpose Specimen Storage Containers
Inert to most chemicals including formaldehyde, weak acids and all bases
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure MaterialColorShape
FKS120899675100 mLLDPEWhiteTall, Thin
FKS12029977118 mLPolypropyleneTranslucentShort, Wide
FIS14-955-115A237 mLLDPETranslucentShort, Wide
FIS14-955-116A237 mLLDPETranslucentTall, Thin
FKS12029957237 mLLDPEWhiteTall, Thin
FIS14-955-117A473 mLLDPETranslucentShort, Wide
FKS11994375473 mLLDPEWhiteTall, Thin
FIS14-955-118A946 mLLDPETranslucentTall, Thin
FIS14-955-120A2500 mLLDPETranslucentTall, Thin
FIS14-955-121A5100 mLLDPETranslucentShort, Wide
FIS14-955-121B5100 mLLDPEWhiteTall, Thin
Catalog number FKS12089967
Price (SGD)
44.00
PK
Add to cart
Capacity (Metric):
5100 mL
Closure Material:
LDPE
Color:
White
Shape:
Tall, Thin
Price (SGD)
44.00
PK
Add to cart
Fisherbrand™ Multipurpose Specimen Containers are designed for collection, transport, and storage of liquid and dry specimens and samples; nonsterile. Inert to most chemicals including formaldehyde, weak acids and all bases.
  • Resist freezing and boiling
  • Stackable for compact storage
  • Select containers are autoclavable (see ordering table); lids are not autoclavable
Specifications
Product TypeSpecimen Container
MaterialHDPE
ShapeTall, Thin
Capacity (English)172 oz.
Capacity (Metric)5100 mL
Closure MaterialLDPE
ColorWhite
Diameter (Metric)21.79 cm
Height (English)6.99 in.
Height (Metric)17.75 cm
Unit SizePK