Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) |
---|---|
FKS12084777 | 5000 mL |
FKS11987564 | 100 mL |
FKS11997564 | 250 mL |
FKS11917574 | 1000 mL |
Measure liquids on bench top or hot plate without fear of chemical attack. Pyrex™ low-form Griffin beakers come in a wide variety of sizes with white gradations and marking spot.