Azlon™ LDPE Round Bottles with Spouted Dropper Closure
Azlon™ LDPE Round Bottles with Spouted Dropper Closure
Actual product may vary
Fisherbrand
Azlon™ LDPE Round Bottles with Spouted Dropper Closure
Use as a dropping or dispensing bottle. Azlon™ LDPE Round Bottles with Spouted Dropper Closure feature a captive sealing cap and can be cut to adjust flow rate.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS12084747150 mL
FKS1070938560 mL
Catalog number FKS12084747
Price (SGD)
78.00
PK
Add to cart
Capacity (Metric):
150 mL
Price (SGD)
78.00
PK
Add to cart
Use as a dropping or dispensing bottle. AzlonTM LDPE Round Bottles with Spouted Dropper Closure feature a captive sealing cap and can be cut to adjust flow rate.
  • Round LDPE bottles fitted with spouted dropper closure
  • Good resistance to chemical attack
Specifications
Capacity (Metric)150 mL
Capacity (English)5.07 oz.
ColorClear
For Use With (Application)Used as a dropping or dispensing bottle
IncludesSpouted dropper closure
MaterialLDPE
QuantityShrink-wrapped pack
Closure IncludedYes
GraduatedNo
MouthNarrow, Threaded
ShapeRound
Height (Metric)147 mm
Product TypeDispensing Bottle
For Use With (Equipment)Used as a Dropping or Dispensing Bottle
Height (English)5.79 in.
Closure MaterialPolypropylene
Closure Size28/R4
Diameter (Metric)50 mm
Closure TypeSpouted Screw Cap
Through the Closure DispensingYes
Unit SizePK