PMP Autoclavable Tweezer
PMP Autoclavable Tweezer
Actual product may vary
Fisher Catalogue
PMP Autoclavable Tweezer
Manufactured with PMP which has a high melting point and good temperature stability. Azlon™ PMP Autoclavable Tweezers are ideal for secure handling of samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
FKS1208458814.5 cm
FKS1205461811.5 cm
Catalog number FKS12084588
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
14.5 cm
Manufactured with PMP which has a high melting point and good temperature stability. Azlon™ PMP Autoclavable Tweezers are ideal for secure handling of samples.
  • High quality, autoclavable tweezers
  • Sharp tip
  • Fine tooth grips for secure handling of samples
Specifications
Tip StyleSharp
MaterialPMP
Length (Metric)14.5 cm
Product TypeTweezers
For Use With (Application)For secure handling of samples
Dimensions (L x W)145 L mm
QuantityEa.
Unit SizePK