Pyrex™ Conical Glass Funnel with PTFE Key
Pyrex™ Conical Glass Funnel with PTFE Key
Actual product may vary
Fisherbrand
Pyrex™ Conical Glass Funnel with PTFE Key
Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ Conical Glass Funnel with interchangeable PTFE stopcock. Complies with ISO 4800 Type 1 Standards.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Joint NumberDescription
FKS12079897250 mL24/29Stopcock Bore: 2.5mm
FKS11373674500 mL24/29Stopcock bore: 4mm
Catalog number FKS12079897
Price (SGD)
409.00
EA
Add to cart
Capacity (Metric):
250 mL
Joint Number:
24/29
Description:
Stopcock Bore: 2.5mm
Price (SGD)
409.00
EA
Add to cart
  • Complies to ISO 4800 Type 1
  • Manufactured from chemically resistant Pyrex™ borosilicate glass
  • With interchangeable PTFE key
  • Supplied with both polyethylene and glass stoppers
Specifications
Capacity (Metric)250 mL
MaterialBorosilicate Glass, Borosilicate Glass
Certifications/ComplianceISO 4800
DescriptionStopcock Bore: 2.5mm
Joint Number24/29
Product TypeSeparating funnel
Unit SizeEA