Pyrex™ Borosilicate Glass Narrow Neck Flat Bottom Boiling Flask
Pyrex™ Borosilicate Glass Narrow Neck Flat Bottom Boiling Flask
Actual product may vary
Fisherbrand
Pyrex™ Borosilicate Glass Narrow Neck Flat Bottom Boiling Flask
Manufactured from Pyrex™ glass for high resistance to thermal shock. Pyrex™ Borosilicate Glass Narrow Neck Flat Bottom Boiling Flask complies with ISO 1773 standards. Excellent chemical durability for contamination-free performance. Includes white enamel marking spot while flat base prevents spillage.
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric) OuterDiameter (Metric) Inner NeckHeight (Metric)
FKS12079002250 mL85 mm30 mm140 mm
Catalog number FKS12079002
Price (SGD)
458.00
PK
Add to cart
Capacity (Metric):
250 mL
Diameter (Metric) Outer:
85 mm
Diameter (Metric) Inner Neck:
30 mm
Height (Metric):
140 mm
Price (SGD)
458.00
PK
Add to cart
  • Complies with ISO 1773
  • Flat base for standing on benches and hotplates etc
  • Manufactured from Pyrex™ glass for high resistance to thermal shock
  • Excellent chemical durability for contamination free performance
  • With white enamel marking spot
Specifications
Certifications/ComplianceISO 1773
Diameter (Metric) Inner Neck30 mm
Capacity (Metric)250 mL
MaterialBorosilicate Glass, Borosilicate Glass
Height (Metric)140 mm
Neck StyleNarrow
Bottom ShapeFlat
Diameter (Metric) Outer85 mm
Unit SizePK