Search Thermo Fisher Scientific
Catalog Number | Cone Size |
---|---|
FKS12078493 | 19/26 (Condenser), 19/26 Fflask) |
Recieve distillate from a condenser. The QuickFit™ distillation splash head reduces the risk of raw liquid entering into the vapor stream.