Polystyrene Petri Dishes
Product Image
Fisherbrand
Polystyrene Petri Dishes
Culture microorganisms on solid media using Thermo Scientific™ Sterilin™ Standard 90mm Petri Dishes. These shallow dishes maximize incubator space and feature a mirror finish that ensures high optical quality.
Have Questions?
Catalog NumberQuantity
FKS12077572600 Case
Catalog number FKS12077572
Price (SGD)
298.00
PK
Add to cart
Quantity:
600 Case
Price (SGD)
298.00
PK
Add to cart
  • Manufactured from non-cytotoxic virgin polystyrene
  • Ideal for use in automatic plate pourers
  • For in vitro use only
  • Triple vented—aids gaseous exchange; ideally suited for short-term work
Specifications
MaterialPolystyrene
ShapeRound
SterilityAseptically Manufactured
TypePetri Dish
Quantity600 Case
Vent3
Height (Metric)14.1 mm
Diameter (Metric)90 mm
Unit SizePK