Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone
Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone
Actual product may vary
Fisherbrand
Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone
Circulates coolant through a coil with a very large cooling surface. Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone is ideal for use in the upright reflux mode where counter-current cooling is possible. Supplied with detachable plastic connectors for the attachment of rubber hose.
Have Questions?
Catalog NumberLength (Metric) JacketStandard Taper Joint No
FKS12070063380 mm (Overall) 207 mm (Effective)40/38
Catalog number FKS12070063
Price (SGD)
476.00
EA
Add to cart
Length (Metric) Jacket:
380 mm (Overall) 207 mm (Effective)
Standard Taper Joint No:
40/38
Price (SGD)
476.00
EA
Add to cart
Circulates coolant through a coil with a very large cooling surface. Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone is ideal for use in the upright reflux mode where counter-current cooling is possible. Supplied with detachable plastic connectors for the attachment of rubber hose.
  • Coolant is circulated through a coil having a very large cooling surface 
  • Ideal for use in the upright reflux mode where counter-current cooling is possible
  • Supplied with detachable plastic connectors for the attachment of rubber hose
Specifications
For Use With (Application)Ideal for use in the upright reflux mode where counter-current cooling is possible
Length (Metric) Jacket380 mm (Overall) 207 mm (Effective)
Standard Taper Joint No40/38
MaterialBorosilicate Glass
TypeCondenser Coil
Unit SizeEA