Dispolab™ Sample Cup Stoppers
Dispolab™ Sample Cup Stoppers
Actual product may vary
Fisher Catalogue
Dispolab™ Sample Cup Stoppers
Available in the push-in design.
Have Questions?
Catalog NumberDiameter (Metric)
FKS1206990716mm
Catalog number FKS12069907
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
16mm
Available in the push-in design.
  • Made from polyethylene
  • For use with Centrifichem™, Technicon™ and Beckman™ analyzers
Specifications
DescriptionPush-on
Diameter (Metric)16mm
MaterialPolyethylene
Product TypeSample Cup Stopper
Unit SizePK