Check Order Status
Disposal Bins
Actual product may vary
Fisherbrand
Disposal Bins
For safe disposal of broken glass
Have Questions?
Catalog Number
Height (Metric)
Width (Metric)
FKS12060133
26.5 cm
23 cm
Catalog number
FKS12060133
Price (SGD)
203.00
EA
Add to cart
Height (Metric):
26.5 cm
Width (Metric):
23 cm
Price (SGD)
203.00
EA
Add to cart
For safe disposal of broken glass
Self assembly rigid boxes
Glass is safely held in strong leak proof and puncture resistant LDPE liner
Lid has an integral waste flap for easy use and has a sealing feature for use when full
Boxes are clearly printed with instructions for use
Specifications
Depth (Metric)
23 cm
Color
Translucent
Height (Metric)
26.5 cm
Shape
Rectangular
Width (Metric)
23 cm
For Use With (Application)
Safe Disposal of Broken Glass
Mounting Type
Bench Top
Material
LDPE
Product Type
Disposal Bin