Nalgene™ PETG Biotainer™ Bottles
Nalgene™ PETG Biotainer™ Bottles
Actual product may vary
Fisherbrand
Nalgene™ PETG Biotainer™ Bottles
Eliminate the cost of cleaning, sterilizing and associated validations compared to bottles designed for reuse with Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles, ideal for prolonged storage of pH sensitive materials such as culture media.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Product TypeQuantity
FKS120599871 LMedia Bottle7 x 5 Bottles
FIS02-540-2631 LMedia Bottle7 x 5 Bottles
FKS104405685 LMedia Bottle6 Bottles
FIS02-540-7015 LSquare Bottle with Handle6 Bottles
Catalog number FKS12059987
Price (SGD)
1,743.00
PK
Add to cart
Capacity (Metric):
1 L
Product Type:
Media Bottle
Quantity:
7 x 5 Bottles
Price (SGD)
1,743.00
PK
Add to cart
Eliminate the cost of cleaning, sterilizing and associated validations compared to bottles designed for reuse with Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles, ideal for prolonged storage of pH sensitive materials such as culture media. Certified sterile and ready-to-use, all Biotainer bottles are gamma irradiated to a Sterility Assurance Level (SAL) of 10–6.
  • Durable, crystal clear PETG for sample integrity
  • Closures with polyethylene foam core liners for leakproof† assurance
  • Custom options available for fluid transfer applications
  • Supported by extensive validation documentation
  • Suitable for storage in conditions from -40° to +70°C
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity (Metric)1 L
Capacity (English)33.8 oz.
ColorClear
Diameter (English)3.8 in.
Diameter (English) Inner Neck1.46 in.
Diameter (Metric) Inner Neck37 mm
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​
MaterialPETG
No. per Case35/Cs.
No. per Pack5/Pk.
Quantity7 x 5 Bottles
SterilitySterile
Temperature Range (Metric)-70°C to 70°C
Certifications/ComplianceUSP Class VI, Non-cytotoxic, USP <661>, Non-pyrogenic, FDA 21 CFR 177.1315
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthNarrow
ShapeSquare
Height (Metric)201 mm
Product TypeMedia Bottle
Height (English)7.9 in.
Closure LinerPolyethylene Foam Core
Closure Size48 mm
Diameter (Metric)98 mm
Unit SizePK