Check Order Status
Quickfit™ Plain Bend Delivery Receiver
Actual product may vary
Fisherbrand
Quickfit™ Plain Bend Delivery Receiver
Plain bend without vent
Have Questions?
Catalog Number
Height (Metric)
Cone Size
FKS12059111
125 mm
24/29
Catalog number
FKS12059111
Price (SGD)
173.00
EA
Add to cart
Height (Metric):
125 mm
Cone Size:
24/29
Price (SGD)
173.00
EA
Add to cart
Quickfit™ Plain Bend Delivery Receiver adapters are available in a range of cone sizes.
Plain bend without vent
Specifications
Material
Borosilicate Glass
Height (Metric)
125 mm
Height (English)
4.9 in.
Cone Size
24/29
Product Type
Receiver Adapter