Polypropylene Scoops
Polypropylene Scoops
Actual product may vary
Fisherbrand
Polypropylene Scoops
Polypropylene construction. Includes sturdy integrated handle.
Have Questions?
Catalog NumberCapacity (Metric)
FKS12055292500 mL
Catalog number FKS12055292
Price (SGD)
147.00
PK
Add to cart
Capacity (Metric):
500 mL
Price (SGD)
147.00
PK
Add to cart
  • Polypropylene construction
  • Integrated handle
  • White
Specifications
Product TypeScoop
MaterialPolypropylene
Capacity (Metric)500 mL
ColorWhite
Length (Metric)315 mm
Unit SizePK