Pipette Filler with Adapter
Pipette Filler with Adapter
Actual product may vary
Fisherbrand
Pipette Filler with Adapter
Kartell™ Pipette Filler with Adapter is compatable with bulb or cylindrical pipettes of various capacities and features no metal parts that could potentially corrode.
Have Questions?
Catalog NumberType
FKS12054717Ball
Catalog number FKS12054717
Price (SGD)
37.00
EA
Add to cart
Type:
Ball
Price (SGD)
37.00
EA
Add to cart
Kartell™ Pipette Filler with Adapter is compatable with bulb or cylindrical pipettes of various capacities and features no metal parts that could potentially corrode.
  • Moulded red rubber bulb
  • Included three glass ball valves which can be controlled by finger pressure
  • There are no metal parts to corrode and the filler can be used with all liquids except those that attack rubber
  • Can be used on bulb or cylindrical pipettes of different capacity
  • Complete with adapter for pipettes with capacities more than 20mL
Specifications
ColorRed
MaterialRubber
TypeBall
Volume (Metric)20 mL
IncludesAdapter for pipettes with capacities greater than 20mL
Unit SizeEA