Azlon™ Plastic Imhoff Sedimentation Cone
Azlon™ Plastic Imhoff Sedimentation Cone
Actual product may vary
Fisherbrand
Azlon™ Plastic Imhoff Sedimentation Cone
Colorless cone made from transparent styrene-acrylonitrile S.A.N polymer with moulded graduations
Have Questions?
Catalog NumberVolume (Metric)
FKS120533741000 mL
Catalog number FKS12053374
Price (SGD)
763.00
EA
Add to cart
Volume (Metric):
1000 mL
Price (SGD)
763.00
EA
Add to cart
  • Colorless, transparent S. A. N cone with molded graduations
  • Graduations from 2mL to 1,000mL (in mixed graduations)
  • The bottom end has a stopper (spare included) to facilitate removal of sample and cleaning
Specifications
Certifications/ComplianceDIN 12672
Volume (Metric)1000 mL
For Use With (Application)For determining the amount of sediment in sewage or industrial waste
Graduated2 to 1000 mL
IncludesStopper
MaterialPlastic
Unit SizeEA