Sterilin™ Polystyrene Containers, 60mL to 250mL
Sterilin™ Polystyrene Containers, 60mL to 250mL
Actual product may vary
Fisherbrand
Sterilin™ Polystyrene Containers, 60mL to 250mL
Contain samples in Thermo Scientific™ Sterilin™ Polystyrene Containers with 60mL to 250mL capacities. Available with either plastic or metal flow seal caps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure MaterialLabelNo. per Case
FKS12047492250 mLMetalPlain50/Cs.
FKS1133915360 mLMetalPlain300/Cs.
FKS1138912360 mLPolyethyleneNo Label300/Cs.
FKS1178889860 mLPolyethylenePrinted300/Cs.
FKS1170890860 mLPolyethylenePlain300/Cs.
FKS11309133100 mLPolyethyleneNo Label200/Cs.
FKS11389143150 mLMetalPrinted120/Cs.
FKS11399143150 mLMetalPlain120/Cs.
Catalog number FKS12047492
Price (SGD)
464.00
PK
Add to cart
Capacity (Metric):
250 mL
Closure Material:
Metal
Label:
Plain
No. per Case:
50/Cs.
Price (SGD)
464.00
PK
Add to cart
Contain samples in Thermo Scientific™ Sterilin™ Polystyrene Containers with 60mL to 250mL capacities. Available with either plastic or metal flow seal caps.
NOT AVAILABLE IN AMERICA
Features:
  • Leak-tested in accordance with BS EN ISO 6717
Ordering Information:
Product is not available in North America.
Specifications
Product TypeContainer
Closure IncludedYes
MaterialPolystyrene
ShapeCylindrical
SterilityNon-sterile
Capacity (Metric)250 mL
Certifications/ComplianceLeak-tested in accordance with BS EN ISO 6717
Closure MaterialMetal
Diameter (Metric)65.5 mm
Height (Metric)119.5 mm
LabelPlain
No. per Case50/Cs.
Unit SizePK