Azlon™ LDPE Wide Neck Wash Bottles
Azlon™ LDPE Wide Neck Wash Bottles
Actual product may vary
Fisher Catalogue
Azlon™ LDPE Wide Neck Wash Bottles
Use these translucent plastic bottles for a variety of liquid storage and sampling applications. Azlon™ LDPE Wide Neck Wash Bottles come with color-coded one-piece PP closures that maintain their shape in use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Color
FKS12044857250 mLYellow
FKS12039847250 mLBlue
FKS120348271000 mLWhite
FKS104422151000 mLGreen
FKS120446971000 mLYellow
FKS101860641000 mLBlue
FKS107076551000 mLRed
Catalog number FKS12044857
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
250 mL
Closure Color:
Yellow
Use these translucent plastic bottles for a variety of liquid storage and sampling applications. AzlonTM LDPE Wide Neck Wash Bottles come with color-coded one-piece PP closures that maintain their shape in use.
  • Square shoulder wash bottles featuring a wide neck for easy spill free filling
  • Fitted with color coded one-piece PP closures that maintain their shape in use
  • Closures feature precision jet-tip aperture for fine control
Specifications
Capacity (Metric)250 mL
Capacity (English)8.45 oz.
ColorTranslucent
IncludesYellow Polypropylene closure
MaterialLDPE
Quantity5/PK
Temperature Range (Metric)–50 to 80°C
Closure ColorYellow
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthWide
ShapeRound
Height (Metric)140 mm
Product TypeWash Bottle
Width (English)2.36 in.
Height (English)5.51 in.
Closure MaterialPolypropylene
Diameter (Metric)60 mm
Closure TypeCap
Through the Closure DispensingYes
Unit SizePK