Pyrex™ Borosilicate Glass Petri Dishes
Pyrex™ Borosilicate Glass Petri Dishes
Actual product may vary
Fisher Catalogue
Pyrex™ Borosilicate Glass Petri Dishes
Manufactured from Pyrex™ borosilicate glass for superior chemical resistance and clarity. Pyrex™ Borosilicate Glass Petri Dishes are a reliable choice for growing bacteria or fungi under laboratory conditions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Height (Metric)
FKS12043333114 mm20 mm
FKS1201333355 mm20 mm
FKS1202333375 mm20 mm
FKS11333704143 mm30 mm
Catalog number FKS12043333
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
114 mm
Height (Metric):
20 mm
  • Specially formed to ensure even wall thickness and consistent optical performance
  • Autoclavable
  • Tight fitting lid safeguards clean environment
Specifications
Closure IncludedClosure Included
Closure MaterialBorosilicate Glass
MaterialBorosilicate Glass
ShapeRound
TypePetri Dish
Height (Metric)20 mm
Diameter (Metric)114 mm
Unit SizePK