Neoprene O-rings
Neoprene O-rings
Actual product may vary
Fisherbrand
Neoprene O-rings
Complete a filtration and vacuum desiccator with a spare o-ring. Kartell™ Neoprene O-rings form a seal with the transparent cover and polypropylene base of a filtration and vacuum desiccator.
Have Questions?
Catalog NumberFor Use With (Equipment)
FKS12034707For 230
Catalog number FKS12034707
Price (SGD)
18.00
EA
Add to cart
For Use With (Equipment):
For 230
Price (SGD)
18.00
EA
Add to cart
  • Essectial component of a Filtration and Vacuum Desiccator
  • Spare O-ring
  • Spread a little grease on O-ring for assistance in establishing initial vacuum
Specifications
ColorBlack
For Use With (Equipment)For 230
MaterialNeoprene
Quantity1Each
Product TypeO-Ring
Unit SizeEA