Quickfit™ Three-Neck Round-Bottom Glass Flask
Quickfit™ Three-Neck Round-Bottom Glass Flask
Actual product may vary
Fisherbrand
Quickfit™ Three-Neck Round-Bottom Glass Flask
Useful for introducing gas inlet tubes and thermometers into the flask. The Quickfit Jointed Ware™ Quickfit™ Three-Neck Round-Bottom Flask has precision-ground sockets from borosilicate glass.
Have Questions?
Catalog NumberCapacity (Metric)Socket Joint
FKS12030673500 mL24/29 (Centre), 19/26 (side)
Catalog number FKS12030673
Price (SGD)
344.00
EA
Add to cart
Capacity (Metric):
500 mL
Socket Joint:
24/29 (Centre), 19/26 (side)
Price (SGD)
344.00
EA
Add to cart
Useful for introducing gas inlet tubes and thermometers into the flask. The Quickfit Jointed Ware™ Quickfit™ Three-Neck Round-Bottom Flask has precision-ground sockets from borosilicate glass.
  • Fitted with three Quickfit™ precision-ground sockets
    • One vertical centre neck
    • Two angled side necks
  • Ground glass sockets are manufactured to an accuracy which surpasses the requirements of ISO 383
  • Side neck useful for introducing gas inlet tubes and thermometers etc., into the flask
  • Manufactured from borosilicate glass for highest resistance to thermal shock and chemical attack
Specifications
Certifications/ComplianceISO 383
Capacity (Metric)500 mL
MaterialBorosilicate Glass
Height (Metric)150 mm
No. per Case3
Socket Joint24/29 (Centre), 19/26 (side)
Bottom ShapeRound
Unit SizeEA