Bottle Brushes
Bottle Brushes
Actual product may vary
Fisher Catalogue
Bottle Brushes
Clean all types of bottles and tubes. B2M™ Bottle Brushes are available with silk, nylon or goat hair bristles for the full range of uses. Brush styles and sizes are available for efficient cleaning of any size or type of receptacle.
Have Questions?
Catalog NumberLength (Metric) HandleLength (Metric) BrushDiameter (Metric) BristleTip Type
FKS12028768500 mm100 mm13 mmReturn End
Catalog number FKS12028768
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric) Handle:
500 mm
Length (Metric) Brush:
100 mm
Diameter (Metric) Bristle:
13 mm
Tip Type:
Return End
  • Three choices of bristles:
    • White silk bristles for general cleaning use
    • Nylon bristles are very resistant for use with aggressive substances
    • Goat hair bristles are very soft for delicate cleaning
  • A variety of tip styles, including: bound-bristle, 2 wicks, cotton and shell tips
  • For cleaning all receptacles: tubes, burettes, balls and bottles
Specifications
Product TypeBottle Brush
Bristle MaterialNylon
Tip TypeReturn End
Diameter (Metric) Bristle13 mm
For Use With (Application)General purpose cleaning, With aggressive substances
For Use With (Equipment)Test Tube
Length (Metric) Brush100 mm
Length (Metric) Handle500 mm
Quantity10 Pk.
Unit SizePK