MBL™ Borosilicate Glass Class A Volumetric Flask with Stopper
MBL™ Borosilicate Glass Class A Volumetric Flask with Stopper
Actual product may vary
MBL™ Borosilicate Glass Class A Volumetric Flask with Stopper
MBL™ Borosilicate Glass Class A Volumetric Flask with Stopper
Fisher Catalogue
MBL™ Borosilicate Glass Class A Volumetric Flask with Stopper
Complies with ISO 1042 and DIN 12664. MBL™ Borosilicate Glass Class A Volumetric Flask with Stopper are manufactured from chemically resistant borosilicate glass.
Have Questions?
Catalog NumberCapacity (Metric)Closure SizeHeight (Metric)
FKS120107331000 mL24/29300 mm
Catalog number FKS12010733
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Closure Size:
24/29
Height (Metric):
300 mm
  • Finely calibrated to Class A tolerances
  • Each flask is laser etched with an individual serial number, batch code and date of manufacture
  • Conformity 'H' certified
  • Inscriptions in blue enamel
  • Supplied with polyethylene stopper
  • Batch certificate available to download online
Specifications
Certifications/ComplianceClass A, ISO 1042, DIN 12664, Conformity 'H' certified
Closure MaterialPolyethylene
DescriptionBlue enamel inscriptions
Capacity (Metric)1000 mL
MaterialBorosilicate Glass
Height (Metric)300 mm
Tolerance±0.4 mL
GraduatedYes
ClassA
Closure Size24/29
Unit SizePK