Use this adapter with the Bibyjet Pro Pipette Controller. Stuart™ Pipette Controller Adapters are specially designed to provide a tight fit for all glass or plastic pipettes from 0.1 to 100mL.
Have Questions?
Catalog Number
For Use With (Equipment)
FKS12008681
Bibbyjet Pro Pipette Controller PC2000/2001
Catalog number FKS12008681
Price (SGD)
173.00
EA
Add to cart
For Use With (Equipment):
Bibbyjet Pro Pipette Controller PC2000/2001
Price (SGD)
173.00
EA
Add to cart
Silicone Adapter
For use with PC2000 Bibbyjet Pro Pipette Controller
Lightweight and ergonomic
Powerful enough to fill a 50mL pipette in just 10s
Battery sufficient to give a full day's work before recharging - LED display shows when battery is low
Pipette holder fully autoclavable
Venting feature allows air sucked from the pipette to be discharged from the front of the unit, avoiding corrosion of internal mechanisms
Specifications
AutoclavableAutoclavable
MaterialSilicone
TypePipette Controller Adapter
For Use With (Equipment)Bibbyjet Pro Pipette Controller PC2000/2001