Azlon™ Retort
Azlon™ Retort
Actual product may vary
Fisherbrand
Azlon™ Retort
Polyethylene coated metal rod screws into the base.
Have Questions?
Catalog NumberDescription
FKS12003374Rod Position: Edge
Catalog number FKS12003374
Price (SGD)
931.00
EA
Add to cart
Description:
Rod Position: Edge
Price (SGD)
931.00
EA
Add to cart
Polyethylene coated metal rod screws into the base.
  • Weighs 2.5kg ensuring stability
  • Corrosion resistant
  • Available with the hole in the centre or at the short edge
Specifications
Dimensions (L x W x H)171 x 248 x 52 mm (Base), 12.5 x 760 mm (Rod)
Product TypeRetort
DescriptionRod Position: Edge
Quantity1/Pk.
Unit SizeEA