Quickfit™ Multiple Glass Joint Adapter with Two Parallel Necks
Quickfit™ Multiple Glass Joint Adapter with Two Parallel Necks
Actual product may vary
Fisherbrand
Quickfit™ Multiple Glass Joint Adapter with Two Parallel Necks
Connect two apparatus together leak-tight. The QuickFit™ multiple glass joint adapter with two parallel necks is the perfect choice.
Have Questions?
Catalog NumberCone SizeStandard Taper Joint No.
FKS1200160314/2314/23
Catalog number FKS12001603
Price (SGD)
247.00
EA
Add to cart
Cone Size:
14/23
Standard Taper Joint No.:
14/23
Price (SGD)
247.00
EA
Add to cart
Connect two apparatus together leak-tight. The QuickFit™ multiple glass joint adapter with two parallel necks is the perfect choice.
With two parallel necks
Specifications
Cone Size14/23
MaterialBorosilicate Glass
Product TypeMultiple neck adapter
Standard Taper Joint No.14/23
Unit SizeEA