Solid Polyethylene Acid Cabinets: Two-Door Stand-Alone Model
Solid Polyethylene Acid Cabinets: Two-Door Stand-Alone Model
Actual product may vary
Solid Polyethylene Acid Cabinets: Two-Door Stand-Alone Model
Solid Polyethylene Acid Cabinets: Two-Door Stand-Alone Model
Fisherbrand
Solid Polyethylene Acid Cabinets: Two-Door Stand-Alone Model
Superior protection against harsh acids, rust and corrosion
Have Questions?
Catalog NumberDoor Style
FKS11999375Manual-close
Catalog number FKS11999375
Price (SGD)
-
Door Style:
Manual-close
Features protective, all-welded bottom sump and a clean, sleek design to complement laboratory settings
  • Sturdy solid-wall, chemically resistant polyethylene construction with sealed sump
  • Suitable for highly aggressive liquids such as phenol, nitric acid, and sulfuric acid
  • 3-point door latch on undercounter and stand alone model offers secure closure and vapor control
  • External 5.08cm (2 in.) NPT vent/exhaust connections
  • Padlockable hasps
  • Sturdy adjustable shelf
  • Leveling feet on all four corners
Specifications
Depth (English) Interior20.87 in.
Depth (Metric) Exterior55.2 cm
Depth (Metric) Interior53 cm
Dimensions (D x W x H) Interior20.87 x 35.5 x 35.5 in. (53 x 90.2 x 90.2 cm)
Door StyleManual-close
Height (English) Interior35.5 in.
Height (Metric) Exterior93.3 cm
Height (Metric) Interior90.2 cm
IncludesAdjustable Steel leg levelers and patent pending Haz-Alert™ reflective warning labels in 3 languages
Load Bearing Capacity23.77 gal. (89.9 L)
No. of Doors2
No. of Shelves1
Weight (English)160 lb.
Weight (Metric)73 kg
Width (English) Interior35.5 in.
Width (Metric) Exterior91.44 cm
Width (Metric) Interior90.2 cm
ColorWhite
Depth (English) Exterior21.75 in.
Height (English) Exterior36.75 in.
MaterialPolyethylene
Product TypeStorage Cabinet
Width (English) Exterior36 in.
Unit SizeEA