Minigrip™ Polyethylene Write-on Bags
Minigrip™ Polyethylene Write-on Bags
Actual product may vary
Fisherbrand
Minigrip™ Polyethylene Write-on Bags
Store and send samples and stationary in resealable bags. Bryson Packaging™ Minigrip™ Polyethylene Write-on Bags are clear polyethylene bags with three white bars for labelling. Square and rectangular bags are available in a range of sizes. 100 and 1,000 packs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWidth (Metric)Height (Metric)
FKS11998267230 mm325 mm
FKS1135820357 mm76 mm
Catalog number FKS11998267
Price (SGD)
49.00
PK
Add to cart
Width (Metric):
230 mm
Height (Metric):
325 mm
Price (SGD)
49.00
PK
Add to cart
  • Clear polyethylene bags with Grip Seal resealable strips
  • Three generously sized write-on labelling bars for easy identification and organization
  • Ideal for storing stationery and sending samples
  • Available in a range of sizes to meet storage needs
Specifications
DescriptionResealable Write-On Bag
ColorClear with White Bars
For Use With (Application)Ideal for storing stationery and sending samples
MaterialPolyethylene
Thickness (Metric)50 μm
Quantity100 Pack
Width (Metric)230 mm
Height (Metric)325 mm
Unit SizePK