Narrow-Mouth Field Sample Bottles
Narrow-Mouth Field Sample Bottles
Actual product may vary
Narrow-Mouth Field Sample Bottles
Narrow-Mouth Field Sample Bottles
Narrow-Mouth Field Sample Bottles
Fisher Catalogue
Narrow-Mouth Field Sample Bottles
Bulk-packaged for convenient liquid sampling, transport, and storage
Have Questions?
Catalog NumberCapacity (Metric)
FKS11997954500 mL
Catalog number FKS11997954
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
Bulk-packaged for convenient liquid sampling, transport, and storage
  • HDPE for excellent chemical resistance
  • Rigid and translucent
  • Ideal for field sampling
Specifications
Capacity17 oz., 500mL
Capacity (Metric)500 mL
Capacity (English)17 oz.
ColorTranslucent
MaterialHDPE
Neck Finish28
Certifications/ComplianceCCD 077; SW-CDC-001501; EN-8-1007-22-0912
Closure IncludedYes
MouthNarrow
Product TypeNarrow Mouth Field Sample Bottle
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Unit SizePK