Acrylic Desiccator Cabinets
Acrylic Desiccator Cabinets
Actual product may vary
Acrylic Desiccator Cabinets
Acrylic Desiccator Cabinets
Acrylic Desiccator Cabinets
Acrylic Desiccator Cabinets
Fisherbrand
Acrylic Desiccator Cabinets
Dust- and moisture-free storage in refrigerators, freezers and on benchtops
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (English)Height (Metric)No. of Shelves
FKS119978647 in.17.8 cm2 removable adjustable and vented shelves
FIS08-642-23B12 in.30.5 cm3 removable adjustable and vented shelves
FKS1190787412 in.30.5 cm3 removable adjustable and vented shelves
FIS08-642-23C18 in.45.7 cm4 Removable Adjustable and Vented
FKS1191787418 in.45.7 cm4 Removable Adjustable and Vented
Catalog number FKS11997864
Price (SGD)
1,348.00
EA
Add to cart
Height (English):
7 in.
Height (Metric):
17.8 cm
No. of Shelves:
2 removable adjustable and vented shelves
Price (SGD)
1,348.00
EA
Add to cart
Dust- and moisture-free storage in refrigerators, freezers and on benchtops
  • Ideal for storing anhydrous and hygroscopic biological and chemical compounds, as well as electronic assemblies and any components requiring controlled storage conditions
  • Neoprene gasket on door ensures tight seal
  • Door is secured with two stainless-steel latches (large size has three latches) and full-height hinge
  • Removable, lipped tray holds solid desiccant
  • Shelves are removable, adjustable and vented to maximize airflow
Specifications
AutoclavableNot Autoclavable
ColorClear
Cabinet StyleFront Loading
MaterialAcrylic
For Use With (Application)Ideal for storing anhydrous and hygroscopic biological and chemical compounds, as well as electronic assemblies and any components requiring controlled storage conditions
IncludesNeoprene gasket, two stainless steel latches (large size has three latches) and full-height hinge, removable solid desiccant lipped tray, vented shelves
No. of Shelves2 removable adjustable and vented shelves
Dimensions (L x W)12 x 12 in. (30.4 x 30.4 cm)
Length (English)12 in.
Length (Metric)30.5 cm
Width (English)12 in.
Width (Metric)30.5 cm
Height (English)7 in.
Height (Metric)17.8 cm
Unit SizeEA