Legato Syringe Pumps
Legato Syringe Pumps
Actual product may vary
Fisherbrand
Legato Syringe Pumps
User-definable flowrates with selectable target volume or time values to control the total infusion volume.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
FKS11995576Legato 210 Syringe Pump, Programmable
FIS10-989-000Legato 200 Syringe Pump
Catalog number FKS11995576
Price (SGD)
28,562.00
PK
Add to cart
Type:
Legato 210 Syringe Pump, Programmable
Price (SGD)
28,562.00
PK
Add to cart
  • Holds one or two syringes from 0.5 μL to 140 mL
  • 6400 microsteps per one revolution of lead screws
  • High resolution color touchscreen
  • Real-time clock
  • Unparalleled ease of use
  • Touch pad “lock” feature
  • LED light on front panel
  • Full metal chasis
  • Built-in syringe table
  • Up to 75 lb. linear force
  • Advanced microstepping techniques
  • Built-in RS-485 interface
  • USB port
  • I/O and TTL interface
  • Continuous mode of operation
  • Spill dam
Specifications
Voltage100/240 V
Flow Rate5 to 220.97 mL/min.
Capacity (Metric)0.5 μL to 140 mL
Certifications/ComplianceCE, UL, CSA, WEE, CB scheme, EU RoHS
ConnectorsRS232 - 9 pin D-sub connector, RS485 - IEEE-1394 6 pos, USB - Type B, I/O and TTL - 15 pin D-sub connector
ControllerMicroprocessor with 0.062 microstepping
Linear Force (Metric)34 kg
Linear Force (English)75 lb.
MaterialSteel
Reproducibility±0.05%
Temperature (Metric) Storage-10°C to +70°C
TypeLegato 210 Syringe Pump, Programmable
Weight (English)10.45 lb.
Weight (Metric)5 kg
Dimensions (L x W x H) Exterior3.5 x 10 x 11 in. (8.89 x 25.4 x 27.94 cm)
Electrical Requirements100/240 V 50/60 Hz
Display TypeTFT
Flow Rate Accuracy±0.35%
Number of Channels2
Frequency50/60 Hz
Overall Height (English)11 in.
Overall Height (Metric)28 cm
Overall Length (English)3.5 in.
Overall Length (Metric)9 cm
Overall Width (English)10 in.
Overall Width (Metric)25.4 cm
Temperature (Metric) Max40°C
Temperature (Metric) Min.4°C
Unit SizePK